Order Entry
Switzerland
ContactUsLinkComponent
Picric acid 13 g/l saturated solution in water, Sigma-Aldrich®
Picric acid 13 g/l saturated solution in water, Sigma-Aldrich®
Katalognr. P6744-1GA
CAS Number:  
undefined
Picric acid 13 g/l saturated solution in water, Sigma-Aldrich®
Katalognr. P6744-1GA
CAS Number:  

Specifications

  • Pk:
    3,78 L
  • Seq:
    5134

Specifications

About this item

 :
 :229.11 g/mol
Merck Index:13,07492
 :M-Clarity™ quality level = MQ200
EINECS:201-865-9
CAS Number:88-89-1
Formula:C₆H₃N₃O₇
MDL Number:MFCD00007102