ContactUsLinkComponent
Velostat® Round Container
Velostat® Round Container
Catalog # DESC57016
Supplier:  DESCO
Velostat® Round Container
Catalog # DESC57016
Supplier:  DESCO
Specifications
  • Description:
    Velostat® round container, size: 60,3×22,2 mm
  • Pk:
    1
Specifications
About this item
Round Velostat® container ideal for static-safe storage, kitting, and in-process handling and transporting of static-sensitive assemblies and devices.
  • Black, injected molded container
  • Lid included