2-Chloroethanol
2-Chloroethanol, Neat, 80.51, 100 mg
ACSDSB442.100MG
: CPACHEM
- Pk:100 mg
- Description:2-Chloroethanol
- Molecular weight (Mw):80.51
- Form:Neat
- Cat. No.:ACSDSB442.100MG
- Formula:C₂H₅ClO
- Storage temperature:4 °C
- CAS No.:107-07-3
- Shipping conditions:20 °C, Air excepted air forbidden



