Hei-PLATE Sensor Packages Expert
HEID506-31151-00
- Description:Hei-PLATE sensor advanced package expert
- Seq:02
- Environmentally Preferable:
- Pk:1
Hei-PLATE sensor Basic and Advanced package Expert.
Basic package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, and Pt1000 temperature sensor for precise temperature regulation.
Advanced package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, Pt1000 temperature sensor for precise temperature regulation, Pt1000 holder with cable device for securely attaching the temperature sensor and silicone protective cover for a longer lasting protection of your magnetic stirrer.