VWR® Jar for Anaerobic Cultures
Supplier: VWR Chemicals
Certificates
About this item
This jar is designed for anaerobic, microaerophilic and or different concentrations of CO₂ cultures. It can be used with any sachet to generate special atmospheres and to contain up to 12 plates.
- No catalyst required
- Metal cover subjected by a threaded closure pressure
- NBR O-ring
Two colours of lid available (red or blue) to easily identify different incubation atmospheres inside the jars during use.