Chucks for RZR Series Electronic Stirrers
Chucks are designed for use with all RZR series overhead electronic stirrers.
The chuck and key set features a hard chrome finish. The flexible shaft features a maximum torque of 150 Ncm.
Frequently Bought Together