Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Lab Jack E-Lift

185×350 mm, 10 kg, 95 - 275 mm
  HEID509-00100-00
Lab Jack E-Lift
  HEID509-00100-00
Lab Jack E-Lift

185×350 mm, 10 kg, 95 - 275 mm

 
SFr. 2’070.00
   
1 * 1 ST
 

Comfortable and secure work with the electric lab jack e-Lift with adjusted plate (190×400 mm) as the magnetic stirrer does not wobble or slip.

  • Power supply via magnetic stirrer/USB-C cable
  • Footprint (W×D): 185×390 mm, including handwheel
  • Compatible with the Hei-PLATE Mix 'n' Heat Ultimate magnetic stirrer

A winch for manual operation and an attachment for additional support rods on the lab jack are both included - for a secure fixing of your setups. The height can be adjusted.

 : Protection class (EN 60529) Mechanic IP 02, electronics IP 42.

 
  • Max. static load capacity:
    10 kg
  • Plate size:
    185×350 mm
  • Working height:
    95 - 275 mm
Accessories

 

  •  
    Description
    For
     
     
  • HEID509-00130-00
    e-Lift accessories set, includes a support rod extension (250 mm) for the adaptation of temperature sensors (for example) as well as an adjustable end position for limiting the travel distance
    e-Lift lab jack
       
    1 * 1 ST - SFr. 269.00