Supelco® BIOshell™ Columns, 160 Å Peptide C18 Fused-Core®
Nome del fornitore: Merck
Certificati
Informazioni su questo articolo
BIOshell™ 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.
- The sterically-protected C18 lighands provide extra stability
- Higher resolutions and higher peak capacities of peptides at ~50% backpressures of sub-2 µm fully porous paticles (FPP)-packed columns
- Lower backpressure allows for columns to be used in series to maximize resolutions and peak capacity of complex proteomic samples or tryptic digests
- Compatible with all UHPLC and LC-MS instrumentation
- Column can be used at high temperatues (up to 90 °C)
The columns are specifically engineered to provide efficient separation of peptides as well as small proteins. These columns contain advanced Fuse-Core® particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. This geatly expands the application range for the separation of biomolecules.
Certifications: Column test certificate available inside the box.
Accessories information: This guard cartridge holder can be used with 4,6 mm/3,0 mm and 2,1 mm Int.Ø BIOShell™ columns.
Specifiche
- Catalog No:
- 67015-U
- 67016-U
- 67099-U
- 67013-U
- 67014-U
- 67017-U
- 67006-U
- 67093-U
- 67282-U
- 67097-U
- 67011-U
- 67012-U
- 67007-U
- 67096-U
- 67008-U
- 66908-U
- 66909-U
- 66906-U
- 66907-U
- 66913-U
- 67018-U
- 66901-U
- 66904-U
- 66905-U
- 66902-U
- 66903-U
- 67284-U
- 67234-U
- 67238-U
- 67239-U
- 67283-U
- 66915-U
- 66921-U
- 66918-U
- 66919-U
- 66923-U
- 66922-U
- 67277-U
- 67001-U
- 67002-U
- 67281-U
- 67274-U
- 67275-U
- 67003-U
- 67004-U
- 67242-U
- 67263-U
- 67267-U
- 67243-U
- 67257-U
- Application:
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
- Carbon load:
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- 4,6%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Surface area:
- 60 m²/g
- 60 m²/g
- 90 m²/g
- 60 m²/g
- 60 m²/g
- 60 m²/g
- 60 m²/g
- 90 m²/g
- 65 m²/g
- 90 m²/g
- 60 m²/g
- 60 m²/g
- 60 m²/g
- 90 m²/g
- 60 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 60 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 65 m²/g
- 60 m²/g
- 60 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 60 m²/g
- 60 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- 65 m²/g
- Temperature limit:
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1