ContactUsLinkComponent
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
  WTWA205210
 :  WTW
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
  WTWA205210
 :  WTW
  • Description:
    Electrolyte for amperometric oxygen sensors, 1 bottle with 50 ml
  • Pk:
    50 mL