Findenser™ Attachment Kit
Catalog # HEID11-300-008-23
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Description:Attachment kit for Findenser and Findenser Mini
- Pk:1 SET
Specifications
About this item
Attachment kit for Findenser™ and Findenser™ Mini. For a secure set up on your hotplate. Take care of a vertical alignement of the elements once set up.
Delivery: Includes stay bar, 2× boss head, 2× retort clamp.