ContactUsLinkComponent
Chemical Standards Kit for Triple Quad Instruments
Chemical Standards Kit for Triple Quad Instruments
Catalog # EKSI4406127
Supplier:  EKSIGENT
CAS Number:  
Chemical Standards Kit for Triple Quad Instruments
Catalog # EKSI4406127
Supplier:  EKSIGENT
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    MS chemical kit 1, low-high concentration PPGs
  • Environmentally Preferable:
Specifications
About this item
Standards chemical kit with low and high concentrations of polypropylene glycol polymers (PPGs).
  • Ensure data consistency - avoid risking the integrity of your workflow
  • Save time and resources
  • Maintain peak performance - troubleshoot your mass spectrometer in minutes
  • Qualify instrument for IQ/OQ/PQ
This standards kit ensures mass spectrometer to turned for optimal performance and delivers reproducible results.
Delivery: This kit includes all solutions necessary for installation and calibration.
Caution: For research use only. Not for use in diagnostic procedures.