Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric acid for ICP-MS
Catalog # ACSDM112.5NP.L1
Supplier: CPACHEM
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Cobalt
- Symbol:Co
Specifications
About this item
Co 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certifications: ISO 17034, ISO 17025, ISO 9001.