Order Entry
Switzerland
ContactUsLinkComponent
Tetraethyl orthosilicate (TEOS), fluid
Flammable
Irritant
Tetraethyl orthosilicate (TEOS), fluid
Catalog # CMPLSB23130.1G
CAS Number:  
Flammable
Irritant
Tetraethyl orthosilicate (TEOS), fluid
Catalog # CMPLSB23130.1G
CAS Number:  

Specifications

  • Pk:
    1.000 mg
  • Seq:
    2865

Specifications

About this item

 :
 :208.33 g/mol
Boiling Pt:166 °C
Melting Pt:–85,5 °C
Density:0,93 g/cm³
Flash Pt:37 °C
UN:1292
ADR:3,III
 :GHS02
 :GHS07
CAS Number:78-10-4
Formula:Si(OC₂H₅)₄
MDL Number:MFCD00009062