perform® Alkaline Laboratory Cleaner, Classic Concentrate Mucasol
SCUE70003439
- Capacity:2 L
- Description:perform® classic concentrate mucasol
- Pack type:Bottle
- Pk:5
perform® classic concentrate mucasol is used for the thorough cleaning of laboratory equipment, technical systems and consumable items and surfaces made of glass, porcelain, plastic, rubber, stainless steel, brass, zinc and copper.
- Alkaline cleaner for powerful cleaning
- High economic efficiency due to low application concentration
- Free of caustic alkalis and chlorine compounds
- Contains tripotassium orthophosphate
This cleaner powerfully removes blood, protein, oil, grease, silicone, tar, wax, tap grease, resin as well as distillation residues.