ContactUsLinkComponent
CryoPlus™ Rack for Cryogenic Vials
CryoPlus™ Rack for Cryogenic Vials
  THBR4000005
CryoPlus™ Rack for Cryogenic Vials
  THBR4000005
  • For:
    CryoPlus 1, 2, 3, 4
  • Description:
    Square rack, vapor phase
  • Pk:
    1
CryoPlus™ series racks are ideal for use with CryoPlus™ LN2 storage systems.
  • Rack includes: cardboard boxes and cardboard dividers for 5 ml vials
  • 6 cardboard boxes with 100 cell divider
  • Holds: 6 boxes/rack