ContactUsLinkComponent
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Supplier:  CPC COLDER
New Product
Certificates
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Supplier:  CPC COLDER
New Product
Specifications
  • Catalog No:
  • CPCCNS6D17006
  • CPCCNS6D22006
  • CPCCNS6D42006
  • Max. operating pressure:
  • Vacuum to 120 psi, 8,3 bar
  • Vacuum to 120 psi, 8,3 bar
  • Operating temperature:
  • 0...71 °C
  • 0...71 °C