ContactUsLinkComponent
Wall Mounted Document Holder (A3 landscape)
Catalog # TEKMOFDH-A3L
Supplier:  TEKNOMEK
Wall Mounted Document Holder (A3 landscape)
Catalog # TEKMOFDH-A3L
Supplier:  TEKNOMEK
Specifications
  • Description:
    Stainless steel A3 landscape wall mounted document holders, 440×280×100 mm
  • Pk:
    1
Specifications
About this item
Fully welded, 304-grade stainless steel A3 landscape wall mounted document holders come in a brushed dull polish finish and have a stackable design.
  • Brushed dull polish finish
  • Wall mounted recipe card/document holders
  • 1 tier stackable
  • Crevice free, easy to clean
Durable and long-lasting solution to document storage needs. Complete with drainage holes for easy cleaning, they are sturdy and hygienic.