Order Entry
Switzerland
ContactUsLinkComponent
Coleman FyreLite Start Gas Stove
  CAMP2203035
Coleman FyreLite Start Gas Stove
  CAMP2203035

 

  • Power:
    3200 W
  • Description:
    Start gas stove, 228 g/h gas consumption, weight: 102 g
  • W×D×H:
    911×430×860 mm
  • Pk:
    1

 

 

The Coleman FyreLite Start gas stove is a good backpacking stove with a good price-performance ratio.

  • Made of high-quality stainless steel
  • Foldable pot racks for compact storage
  • It has a cross over the burner as a windbreak

Ideal for backpackers or campers when weight is an important consideration. The stove weighs 102 g and has an output of 3200 W. Comes with integrated wind protection, the stove delivers high performance, even in light winds.

The stainless steel pot support provides a stable support when cooking and can be easily folded for easy transport when not in use.

The device can be operated with Coleman® Xtreme Gas and Coleman® Performance Gas.

 : A bag (100% polyester) is included in delivery.