Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nickel Standard Solution

Nickel Standard Solution

Nickel standard solution, 1000 mg/L, 500 mL
Catalog # 713-4089
Supplier:   WTW
Nickel Standard Solution
Catalog # 713-4089
Supplier:  WTW
CAS Number:  
Nickel Standard Solution
Nickel Standard Solution

Nickel standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
SFr. 82.70
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Nickel standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.