Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lead Standard Solution

Lead Standard Solution

Lead standard solution, 1000 mg/L, 500 mL
Catalog # 713-4074
Supplier:   WTW
Lead Standard Solution
Catalog # 713-4074
Supplier:  WTW
CAS Number:  
Lead Standard Solution
Lead Standard Solution

Lead standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
SFr. 81.30
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Lead standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.