Order Entry
Switzerland
ContactUsLinkComponent
T4 GP32 Protein (from E.coli)
T4 GP32 Protein (from E.coli)
Catalog # GENO786-1996
Supplier:  G-Biosciences
CAS Number:  
T4 GP32 Protein (from E.coli)
Catalog # GENO786-1996
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Enzyme name:
    T4 gp32 Protein

Specifications

About this item

T4 GP32 is a single-stranded DNA binding protein with key role in DNA replication, recombination and repair. T4 GP32 binds cooperatively to single stranded DNA and stimulates cell-free DNA synthesis directed by the T4-encoded DNA polymerase. It improves the efficiency of reverse transcriptase during RT-PCR as well as increases the yield of PCR products.

  • Product source: E. coli cells expressing T4 gp32 gene
  • Storage temperature: −20 °C
  • Heat inactivation: 65 °C for 20 min
  • Quality control assays: T4 GP32 protein is free from detectable nuclease contamination

Product includes: T4 GP32 protein and 10X GP32 reaction buffer

10X GP 32 reaction buffer composition:
200 mM Tris-acetate
1M Potassium acetate
100 mM Magnesium acetate
10 mM DTT
pH 7,8 at 25 °C