Nunc™ EasYDish™ Cell Culture Dishes with Nunclon™ Supra Surface
Anbieter: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
Experience optimal performance with Thermo Scientific Nunc™ EasYDish™ dishes with Nunclon™ Supra surface treatment for culturing a wide variety of fastidious adherent cells.
- The beveled grip ring is easy to hold and stays secure in your gloved hand
- Available in four sizes: 35, 60, 100, and 150 mm
- Sterile, not autoclavable
- Clear, round, flat bottom, not gridded
- Single use
- Consistent lot-to-lot performance
- Non-cytotoxic and non-pyrogenic
- Biocompatible per USP Class VI for raw materials
- 5 years of shelf life
Intended for general cell culture applications, the Nunclon™ Supra surface is more hydrophilic than the Nunclon™ Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon™ Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon™ Supra cultureware is a great xeno-free solution for cell therapy research applications.
Technische Daten
- Catalog No:
- 734-3639
- 734-3636
- 734-3643
- 734-3638
- 734-3641
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS
- IRECS
- IRECS
- IRECS
- IRECS
Technische Daten
Frequently Bought Together