Water Disinfector, WD6010
SMEGWD6010
: SMEG
- Description:Washer disinfector
- Model:WD6010
- Weight:280 Kg
- Pk:1
WD6010 is a single or double glass drop down door washer-disinfector. washing capacity of up to 12 din baskets with 65 cm width. the drop down door allows the operator to ergonomically load and unload the baskets included in the trolley without having to lift heavy weights. also available in a version with turbo cycles (WD6010T model).