SARS-CoV-2 (COVID-19) Recombinant M Protein (from E. coli)
Catalog # PRSI10-429
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles, store at 2...8 °C for one week, store at −20...−80 °C for twelve months from the date of receipt
- Protein synonyms:Membrane protein|Membrane glycoprotein
- Protein/peptide name:Membrane
- Purity:>90% as determined by SDS-PAGE quantitative densitometry by Coomassie Blue Staining
- Molecular weight:15,84 kDa
- Sequence:Arg101-Gln222
- Formulation:Lyophilised, supplied as lyophilised from PBS, pH 7,5, 0,02 % NLS, please reconstitute the protein with ddH₂O to the concentration with 1 mg/ml
- Shipping temperature:RT