About this item
Eppendorf Tubes® with snap caps are an ideal choice when working with medium-sized sample volumes (0,5 to 5,0 ml). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25000×g), and sample recovery.
- Designed for ease of use and user-friendly handling during sample preparation and processing
- Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions
- Specifically designed to enhance sample recovery, preserving the integrity of samples
- Available in Eppendorf Quality, PCR Clean, Biopur®, Sterile, and Forensic DNA Grade quality for tested purity
Explore the versatility of Eppendorf 5,0 ml tubes, available in both colorless and amber options. Tailor the selection with various certified purities, ensuring precise and reliable sample recoveries. Experience the flexibility these tubes provide, allowing selection of the ideal combination for specific research needs.
Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Snap cap
Material: PP
Diameter: 17 mm
Specifications
- Catalog No:
- 525-0797
- 525-1044
- 525-0795
- 525-0743
- 525-0796
- 525-0671
- EPPE0030119.401
- Autoclavable:
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Max. RCF:
- 25000×g
- 25000×g
- 25000×g
- 25000×g
- 25000×g
- 25000×g
- 25000×g
- Operating temperature range:
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- Volume:
- 5,0 ml
- 5,0 ml
- 5,0 ml
- 5,0 ml
- 5,0 ml
- 5,0 ml
- 5,0 ml
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001