Silica ≥99.8%, Supelco®
Anbieter: Merck
Zertifikate
Zertifikate
Über diesen Artikel
CAS-Nummer:112945-52-5
:
Formel:SiO₂
:60.083 g/mol
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Anbieter: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Anbieter: Merck
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
VWR®, Snap Cap Vials
Anbieter: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR®, Slide Boxes with Lid
Anbieter: VWR Collection
POTASSIUM SUCROSE OCTASULFATE 1 * 500 mg
Anbieter: Merck
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Anbieter: EPPENDORF
Sand, quartz 50-70 mesh, Fluka™
Anbieter: Honeywell Chemicals
DNA loading dye TriTrack 6x 5x1ml 1 * 1 KIT
Anbieter: Thermo Fisher Scientific
Water standards for Karl Fischer titration, Hydranal™
Anbieter: Honeywell Chemicals