Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silica gel C18-RP
Silica gel C18-RP, Supelco®
Silica gel C18-RP, Supelco®
Silica gel C18-RP, Supelco®
Catalog # 97727-U
Supplier:
Merck
CAS Number:
Expand image
Silica gel C18-RP, Supelco®
Catalog # 97727-U
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
CAS Number:
71889-02-6
MDL Number:
MFCD00241275
Product Details