Order Entry
Switzerland
ContactUsLinkComponent
Silica gel, spherical 75-200 μm particle size, Supelco®
Silica gel, spherical 75-200 μm particle size, Supelco®
Catalog # 78991-100G
Supplier:  Merck
CAS Number:  
undefined
Silica gel, spherical 75-200 μm particle size, Supelco®
Catalog # 78991-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:> 1600 °C
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232