Silica gel, Davisil® Grade 12 pore size 150 Å, 60-200 mesh, high purity, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:> 1600 °C
Storage Temperature:Ambient
EINECS:231-545-4
Frequently Bought Together