Silica gel (9385) pore size 60 Å, 230-400 mesh particle size, high purity, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:> 1600 °C
Storage Temperature:Ambient
EINECS:231-545-4
Frequently Bought Together