ContactUsLinkComponent
Cerex® FLORASIL SPE Columns
Cerex® FLORASIL SPE Columns
Catalog # TCAT30213512
Supplier:  Tecan
Cerex® FLORASIL SPE Columns
Catalog # TCAT30213512
Supplier:  Tecan
Specifications
  • Capacity:
    6 ml
  • Description:
    Column FLORASIL
  • Bed weight:
    1000 mg
  • Pk:
    50
Specifications
About this item
Florisil is used for sample preparation (solid phase extraction) of e.g., organic tin compounds and aliphatic carboxylic acids. Florisil is a magnesium silicate.
  • Ion-exchange
  • Liquid handler compatible