Acetic acid ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Acetic acid ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
:64-19-7
:
:H₃CCOOH
:MFCD00036152
:60.05 g/mol
:Ambient
:200-580-7
:2790
:8,
:GHS05
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals