Order Entry
Switzerland
ContactUsLinkComponent
Ammonia solution 2 mol/l (2 N) Reag. Ph. Eur. 1004703
Ammonia solution 2 mol/l (2 N) Reag. Ph. Eur. 1004703
Catalog # 87766.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonia solution 2 mol/l (2 N) Reag. Ph. Eur. 1004703
Catalog # 87766.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :GHS05
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650