Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® Table Windshield for Balances
  VWRI611-5598
undefined
VWR® Table Windshield for Balances
  VWRI611-5598

 

  • Description:
    Table top draft windshield
  • Dimensions:
    Overall (W×D×H): 350×500×370 mm
    Internal (W×D×H): 320×460×360 mm
    Side opening (W×H): 200×330 mm
  • Pk:
    1

 

 

Protect laboratory balances with a removable windshield during measurements.

  • Large windshield with a white metal frame
  • Anti-static acrylic with two side openings

This windshield ensures accurate measurements while safeguarding against environmental changes for Analytical, Semimicro, and especially Microbalances.