


Ammonia solution 10% Reag. Ph. Eur. 1004702
Catalog # REGE1004702
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:35.05 g/mol
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650