Order Entry
Switzerland
ContactUsLinkComponent
Ammonium iron (III) citrate (∼18% Fe), SAFC®
Ammonium iron (III) citrate (∼18% Fe), SAFC®
Catalog # 1.03762.1000
CAS Number:  
Ammonium iron (III) citrate (∼18% Fe), SAFC®
Catalog # 1.03762.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
EINECS:214-686-6
CAS Number:1185-57-5
Formula:C₆H₅+4y)Fe(x)N(y)O₇
MDL Number:MFCD00013099