4,4'-Isopropylidenediphenol
Catalog # WAKO308-07711
Supplier: Wako
CAS Number:
Specifications
- Pk:20 Tests
- Seq:0760
Specifications
About this item
:
:228.29 g/mol
Merck Index:13,01296
Boiling Pt:401 °C (1013 hPa)
Melting Pt:155 °C
Density:1.205 g/cm³ (20 °C)
Flash Pt:227 °C
EINECS:201-245-8
CAS Number:80-05-7
Formula:C₁₅H₁₆O₂
MDL Number:MFCD00002366