Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 4,6-Dinitro-o-cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 4,6-Dinitro-o-cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA13050000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 4,6-Dinitro-o-cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA13050000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    534-52-1
  • Description:
    4,6-Dinitro-o-cresol 100 µg/ml in methanol
  • Formula:
    C₇H₆N₂O₅
  • Description:
    4,6-Dinitro-o-cresol

Specifications

About this item