ContactUsLinkComponent
Spec-Tec™ Stretch M104 Sterile Glove Liners
Spec-Tec™ Stretch M104 Sterile Glove Liners
Supplier:  WELLS LAMONT
Certificates
undefined
Spec-Tec™ Stretch M104 Sterile Glove Liners
Supplier:  WELLS LAMONT
Specifications
  • Catalog No:
  • 112-6618
  • 112-6617
  • 112-6614
  • 112-6615
  • 112-6616
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 1000/ISO 6
  • 1000/ISO 6
  • 1000/ISO 6
  • 1000/ISO 6
  • 1000/ISO 6
  • Sterility:
  • Sterile
  • Sterile
  • Sterile
  • Sterile
  • Sterile