Spec-Tec® Spectra® Fiber M102 Sterile Critical Environment Glove Liners
Supplier: WELLS LAMONT
Certificates
About this item
Sterile and disposable cut-resistant glove liners.
- For use in cleanroom environments
- Cut-resistant
This uniquely thin glove is constructed of 100% HPPE fiber that provides comfort underneath a disposable glove. An EN 388 X241 rating provides level 2 cut resistance. These glove liners are individually wrapped in a sterile package. The M102 is an ideal glove in industries such as pharmaceutical, medical, and organ tissue processing.
Certifications: EN 388 X241.
Packaging: Gloves are packaged in a box of 10 pairs. Each pair is sealed in its own sterile pouch.
Specifications
- Catalog No:
- 112-6613
- 112-6610
- 112-6611
- 112-6612
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile