Packo Biobased Jars
: CURTEC
The new Packo lid with integrated gasket prevents liquids from escaping or spilling out. Leak-tightness is tested by resting pots on the lid and side for a certain period of time thus burdening the gasket.
- Quarter-turn screw lid saves handling time
- Low moisture vapor transmission
- Food safe
- Tamper evident
Closing a Packo jar according to the manual is easy enough by hand. Closing and opening however can be made even easier and more ergonomic with a Lid Locker. This simple tool allows you to create more torque with less effort.
: UN certified (1H2) for solids.
Frequently Bought Together