Order Entry
Switzerland
ContactUsLinkComponent
Fetal Bovine Serum, Regular, USDA Approved Origin
Fetal Bovine Serum, Regular, USDA Approved Origin
Catalog # MDTC35-011-CV
Supplier:  Corning
undefined
Fetal Bovine Serum, Regular, USDA Approved Origin
Catalog # MDTC35-011-CV
Supplier:  Corning

Specifications

  • Description:
    Fetal bovine serum, regular, USDA approved origin (heat inactivated), 500 ml
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    500 mL

Specifications

About this item

Wide variety of fetal bovine serum (FBS) options to satisfy most common applications and is a perfect complement to Corning classical cell culture media. Products are evaluated for quality, performance
and analytical testing, including sterility, mycoplasma, virus testing, total protein, endotoxin and hemoglobin levels.

  • Storage temperature is −40 to −10 °C
Certifications: ISIA, CEP, EDOM certified.