Colour Standards ASTM
Supplier: LGC Standards PROMOCHEM
New Product
Certificates
About this item
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together
VWR®, Viscosity standards
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Hydrochloric acid 25% Reag. Ph. Eur. 1043501
Supplier: VWR Chemicals
Accessories for pH/°C meter, handheld
Supplier: Testo
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
MULTIPLE STANDARD SOL. HIGH VARION ES/2 1 * 1 L
Supplier: WTW ONLINE
ISOPROPYLBENZENE 1 * 500 mg
Supplier: EHRENSTORFER
PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
Mercury Hg - 10 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Supplier: ACTIVE MOTIF
BETAHISTINE DIHYDROCHLORIDE 1 * 50 mg
Supplier: EDQM
TBN STANDARD 10 MG/G KOH 1 * 50 g
Supplier: REAGECON
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
Saccharin
Supplier: EHRENSTORFER
Thiamphenicol, analytical standard, Supelco®
Supplier: Merck
Tepraloxydim-3-Hydroxy-Glutaric Acid 1 * 10 mg
Supplier: EHRENSTORFER
PIGMENT YELLOW 1 1 * 50 mg
Supplier: EHRENSTORFER
Supelco® PESTANAL® Organic Reference Standard, Pirimicarb
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach