Informazioni su questo articolo
Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP68 immersion/ IP69K washdown protection, 316 stainless steel construction and hermetically-sealed load cell.
- 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
- 6 D (LR20) Batteries (Included)
- i-DT61PW indicator
Versatile scales include fast weight display and multiple software modes to meet food, chemical processing, and packaging needs.
150% overload capacity protection, 3-colour check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare, adjustable rubber feet.
316 stainless steel platform and frame, hermetically-sealed stainless steel load cell, IP68/IP69K food grade polycarbonate housing indicator and 316 stainless steel mounting bracket.
Weighing, counting with APW optimisation, percent weighing, checkweighing, checkcounting, dynamic weighing/display hold.
Specifiche
- Catalog No:
- OHAU30608731
- OHAU30608730
- OHAU30608729
- OHAU30608728
- OHAU30612510
- OHAU30612506
- OHAU30612505
- OHAU30608732
- OHAU30608667
- OHAU30612509
- OHAU30612508
- OHAU30612507
- Operating temperature range:
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C