Ultra Safety Cutter
Codice catalogo PRTWKN10BKR
Nome del fornitore: Portwest
Specifiche
- Description:Ultra safety cutter
- Pk:1
Specifiche
Informazioni su questo articolo
Cleverly designed using zinc alloy with safety in mind, this knife for right-handed users has a spring-loaded blade retraction system that responds to any uncontrolled hand movement. With an easy blade change, this knife is a great all-rounder.
- Spring-loaded blade retract system responds to uncontrolled hand movement
- Slider button helps to avoid accidents
- Quick and easy blade change system
- Retail tag aids presentation for retail sales
With an easy blade change, this knife is a great all-round utility tool. Designed for right-handed users, the blade is manufactured using zinc alloy for a durable and great value option.