Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (APC/iFluor® 750)
Supplier: AAT BIOQUEST
Certificates
About this item
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: APC/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB100811G0
- AATB100811G1
- AATB100811G2
- Antigen-Symbol:
- CD8a
- CD8a
- CD8a
- Antigen-Name:
- CD8a antigen
- CD8a antigen
- CD8a antigen
- Antikörper-Typ:
- Primär
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Monoklonal
- Konjugation:
- APC (Allophycocyanin)/iFluor® 750
- APC (Allophycocyanin)/iFluor® 750
- APC (Allophycocyanin)/iFluor® 750
- Klon:
- SK1
- SK1
- SK1
- Reaktivität:
- Human
- Human
- Human
- Wirt:
- Mouse
- Mouse
- Mouse
- ImmunFluoreszenz:
- Yes
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Yes
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Kreuzadsorption:
- No
- No
- No
- Lagertemperatur:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Transporttemperatur:
- 20
- 20
- 20
Specifications
Frequently Bought Together