Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Catalog # PRSI91-846
Supplier:  ProSci Inc.
New Product
CAS Number:  
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Catalog # PRSI91-846
Supplier:  ProSci Inc.
New Product
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Enzyme name:
    Palmitoyl-protein thioesterase 1
  • Enzyme synonyms:
    Palmitoyl-protein thioesterase 1, PPT-1, Palmitoyl-protein hydrolase 1, PPT1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32,3 kD
  • Sequence:
    Asp28-Gly306
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7,5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Palmitoyl-protein thioesterase 1(PPT-1 for short), also known as Palmitoyl-protein hydrolase 1, belongs to the palmitoyl-protein thioesterase family.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

It is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. This enzyme removes thioester-linked fatty acyl groups such as palmitate from modified cysteine residues in proteins or peptides during lysosomal degradation. Defects in PPT1 are the cause of neuronal ceroid lipofuscinosis type 1.