Direct Vent Goggles
Direct vent goggles, Clear
Catalog # PRTWPW20CLR
Supplier: Portwest
Jump to:
Product Details & Documents
Designed to be worn in a tough work environment, these goggles are fitted with a flexible PVC frame for ultra-light and comfortable wear.
- Direct ventilation
- Flexible PVC frame for ultra comfort
- Adjustable elastic headband for a perfect fit
- 99% UV protection
- Retail bag aids presentation for retail sales
Lightweight and comfortable direct ventilation goggles offer excellent side protection, and are individually packaged.
Certifications: ANSI/ISEA Z87+.
Specifications
- Colour:Clear
- Description:Direct vent goggles