ContactUsLinkComponent
Lens Cleaning Wipes
Lens Cleaning Wipes
Catalog # PRTWPA01WHR
Supplier:  Portwest
Lens Cleaning Wipes
Catalog # PRTWPA01WHR
Supplier:  Portwest
Specifications
  • Colour:
    White
  • Description:
    Lens cleaning wipes
  • Pk:
    100
Specifications
About this item
Lens Cleaning Wipes are packaged in a handy dispenser box which is pre-assembled for immediate use.
  • 100 towelettes per box
  • Low-linting construction for minimal contamination
  • Each towelette is individually wrapped
  • Suitable for all Portwest safety eyewear
  • Anti-static
  • No alcohol content
Silicone free wipes are ideal for manufacturing, paint applications, electronics, and glass handling applications where silicone is problematic.
Dispenser box measures 130×200 mm, and holds 100 individually packaged lens cleaning wipes.