ContactUsLinkComponent
Valved Dolomite Light Cup Respirator, FFP3
Valved Dolomite Light Cup Respirator, FFP3
Catalog # PRTWP304WHR
Supplier:  Portwest
Valved Dolomite Light Cup Respirator, FFP3
Catalog # PRTWP304WHR
Supplier:  Portwest
Specifications
  • Description:
    FFP3 valved Dolomite light cup respirator mask
  • Pk:
    10
Specifications
About this item
Disposable respirator with a lightweight cup featuring EVA foam under the adjustable nose bridge for extra comfort.
  • Adjustable nose bar for improved fit and reduced fogging of glasses
  • Foam padding around the nose bridge for additional comfort
  • Ultrasonic welded head straps
  • High performance exhalation valve provides comfort in hot and humid conditions
  • Lightweight cup style design
  • Retail box aids presentation for retail sales
Ultrasonic welded and metal free, the exhalation valve works efficiently to reduce heat inside the mask and provides comfort when used in hot and humid conditions. Dolomite optional clogging test (D) to provide more comfortable breathing and longer lasting filtration performance.
Certifications: CE-CAT III.